AMBH324A50CB Ambeed, Inc.

BH3I-1

  • MDL number MFCD03453544
  • Cas Number : 300817-68-9
  • Molecular Weight : 400.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/300817-68-9.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/300817-68-9.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart