AMBH324A50D4 Ambeed, Inc.

(S)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid

  • MDL number MFCD00042632
  • Cas Number : 13726-76-6
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/13726-76-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13726-76-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart