AMBH324A50DB Ambeed, Inc.

Methyl (E)-3-(3-aminophenyl)acrylate

  • MDL number MFCD20527968
  • Cas Number : 163487-10-3
  • Molecular Weight : 177.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/163487-10-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/163487-10-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart