AMBH324A50E1 Ambeed, Inc.

H-Tyr(3-I)-OH

  • MDL number MFCD00002608
  • Cas Number : 70-78-0
  • Molecular Weight : 307.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/70-78-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/70-78-0.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart