AMBH324A50E3 Ambeed, Inc.

(S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran

  • MDL number MFCD27920793
  • Cas Number : 915095-89-5
  • Molecular Weight : 367.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/915095-89-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/915095-89-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart