AMBH324A50EA Ambeed, Inc.

(S)-1-((S)-2-((tert-Butoxycarbonyl)amino)propanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00134853
  • Cas Number : 33300-72-0
  • Molecular Weight : 286.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/33300-72-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/33300-72-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart