AMBH324A50ED Ambeed, Inc.

Dicyclohexylamine (S)-4-(((allyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)butanoate

  • MDL number MFCD01862280
  • Cas Number : 327156-92-3
  • Molecular Weight : 483.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/327156-92-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/327156-92-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart