AMBH324A50F0 Ambeed, Inc.

tert-Butyl (4R-cis)-6-[(acetyloxy)methyl]-2,2-dimethyl-1,3-dioxane-4-acetate

  • MDL number MFCD08458213
  • Cas Number : 154026-95-6
  • Molecular Weight : 302.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/154026-95-6.html

Safety Information

signalword Warning
pcodes P264-P270-P273-P301+P312-P330
MSDS https://www.ambeed.com/products/154026-95-6.html
pictograms GHS07
hstatements H302-H413
Availability details for
Quantity
  • Availability
  • Add to Cart