AMBH324A50F9 Ambeed, Inc.

KPT-276

  • MDL number MFCD27937049
  • Cas Number : 1421919-75-6
  • Molecular Weight : 426.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1421919-75-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1421919-75-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart