AMBH324A50FC Ambeed, Inc.

(2S,3S)-Diethyl 2,3-dihydroxysuccinate

  • MDL number MFCD00064451
  • Cas Number : 13811-71-7
  • Molecular Weight : 206.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13811-71-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13811-71-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart