AMBH324A5101 Ambeed, Inc.

Quinine Sulfate Hydrate

  • MDL number MFCD29075629
  • Cas Number : 6119-70-6
  • Molecular Weight : 818.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6119-70-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/6119-70-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart