AMBH324A510F Ambeed, Inc.

(R)-tert-Butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate

  • MDL number MFCD00674064
  • Cas Number : 95715-87-0
  • Molecular Weight : 229.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/95715-87-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95715-87-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart