AMBH324A5110 Ambeed, Inc.

(S)-tert-Butyl (1-amino-3-methyl-1-oxobutan-2-yl)carbamate

  • MDL number MFCD00190841
  • Cas Number : 35150-08-4
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/35150-08-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/35150-08-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart