AMBH324A5120 Ambeed, Inc.

Cefoperazone

  • MDL number MFCD00865067
  • Cas Number : 62893-19-0
  • Molecular Weight : 645.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/62893-19-0.html

Safety Information

signalword Danger
pcodes P261-P280-P342+P311
MSDS https://www.ambeed.com/products/62893-19-0.html
pictograms GHS08
hstatements H317-H334
Availability details for
Quantity
  • Availability
  • Add to Cart