AMBH324A512B Ambeed, Inc.

(2S,5R)-Ethyl 5-((benzyloxy)amino)piperidine-2-carboxylate oxalate

  • MDL number N/A
  • Cas Number : 1416134-48-9
  • Molecular Weight : 368.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1416134-48-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1416134-48-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart