AMBH324A5133 Ambeed, Inc.

(3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl) methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid tert-butyl ester

  • MDL number MFCD08460212
  • Cas Number : 355806-00-7
  • Molecular Weight : 537.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/355806-00-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/355806-00-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart