AMBH324A514B Ambeed, Inc.

(R)-5-(Chloromethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one

  • MDL number MFCD22683177
  • Cas Number : 496031-57-3
  • Molecular Weight : 314.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/496031-57-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/496031-57-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart