AMBH324A5150 Ambeed, Inc.

(S)-3-(4-(2-Chloro-5-iodobenzyl)phenoxy)tetrahydrofuran

  • MDL number MFCD28098707
  • Cas Number : 915095-94-2
  • Molecular Weight : 414.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/915095-94-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/915095-94-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart