AMBH324A5153 Ambeed, Inc.

4-(trans-4-Heptylcyclohexyl)benzonitrile

  • MDL number MFCD01941020
  • Cas Number : 61204-03-3
  • Molecular Weight : 283.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/61204-03-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/61204-03-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart