AMBH324A5157 Ambeed, Inc.

Boc-Dap(Boc)-OH.DCHA

  • MDL number MFCD00238311
  • Cas Number : 201472-68-6
  • Molecular Weight : 485.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/201472-68-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201472-68-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart