AMBH324A5168 Ambeed, Inc.

Acepromazine maleate

  • MDL number MFCD00082473
  • Cas Number : 3598-37-6
  • Molecular Weight : 442.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3598-37-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/3598-37-6.html
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart