AMBH324A516E Ambeed, Inc.

Fmoc-N-Me-Nle-OH

  • MDL number MFCD00235878
  • Cas Number : 112883-42-8
  • Molecular Weight : 367.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/112883-42-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112883-42-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart