AMBH324A5178 Ambeed, Inc.

(R)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate 4-methylbenzenesulfonate

  • MDL number MFCD00136658
  • Cas Number : 97984-63-9
  • Molecular Weight : 443.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/97984-63-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/97984-63-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart