AMBH324A5197 Ambeed, Inc.

(S)-Isopropyl 2-amino-3-(4-hydroxyphenyl)propanoate

  • MDL number MFCD00871218
  • Cas Number : 126173-94-2
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/126173-94-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/126173-94-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart