AMBH324A51A0 Ambeed, Inc.

trans-4-Propyl-4'-(2,3,4-trifluorophenyl)-1,1'-bi(cyclohexane)

  • MDL number MFCD23115464
  • Cas Number : 139056-62-5
  • Molecular Weight : 338.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/139056-62-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/139056-62-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart