AMBH324A51A3 Ambeed, Inc.

Aliskiren hemifumarate

  • MDL number MFCD10566724
  • Cas Number : 173334-58-2
  • Molecular Weight : 1219.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/173334-58-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/173334-58-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart