AMBH324A51A4 Ambeed, Inc.

Cardiogreen

  • MDL number MFCD00013078
  • Cas Number : 3599-32-4
  • Molecular Weight : 774.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3599-32-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3599-32-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart