AMBH324A51B7 Ambeed, Inc.

(Z)-Methyl 2-(1-(2-chlorophenyl)-4-((3-(dimethylamino)phenyl)(methylamino)methylene)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl)acetate

  • MDL number MFCD28369464
  • Cas Number : 1820587-57-2
  • Molecular Weight : 426.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1820587-57-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1820587-57-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart