AMBH324A51BA Ambeed, Inc.

Dihydroartemisinin (mixture of a and �isomers)

  • MDL number MFCD28384156
  • Cas Number : 131175-87-6
  • Molecular Weight : 284.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/131175-87-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/131175-87-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart