AMBH324A51C2 Ambeed, Inc.

Fmoc-Trp-OPfp

  • MDL number MFCD00065681
  • Cas Number : 86069-87-6
  • Molecular Weight : 592.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/86069-87-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/86069-87-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart