AMBH324A51C7 Ambeed, Inc.

BIs(2-methyl-2-propanyl) (2S,2'S)-2,2'-[4,4'-biphenyldiylbis(1H-imidazole-4,2-diyl)]di(1-pyrrolidinecarboxylate)

  • MDL number MFCD28404669
  • Cas Number : 1007882-23-6
  • Molecular Weight : 624.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1007882-23-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1007882-23-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart