AMBH324A51CD Ambeed, Inc.

(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

  • MDL number MFCD07357308
  • Cas Number : 116539-55-0
  • Molecular Weight : 171.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/116539-55-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/116539-55-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart