AMBH324A51D6 Ambeed, Inc.

(4S,4'S)-2,2'-(Cyclopentane-1,1-diyl)bis(4-benzyl-4,5-dihydrooxazole)

  • MDL number MFCD28015909
  • Cas Number : 1003886-05-2
  • Molecular Weight : 388.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1003886-05-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1003886-05-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart