AMBH324A51E1 Ambeed, Inc.

Z-D-Arg-OH.HCl

  • MDL number MFCD00063010
  • Cas Number : 113712-05-3
  • Molecular Weight : 344.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/113712-05-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/113712-05-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart