AMBH324A51E7 Ambeed, Inc.

(R)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate acetate

  • MDL number MFCD30475785
  • Cas Number : 1998701-09-9
  • Molecular Weight : 276.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1998701-09-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1998701-09-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart