AMBH324A51E8 Ambeed, Inc.

(S)-Diethyl 2-(5-(methylamino)thiophene-2-carboxamido)pentanedioate

  • MDL number MFCD09750956
  • Cas Number : 112889-02-8
  • Molecular Weight : 342.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/112889-02-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112889-02-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart