AMBH324A51F4 Ambeed, Inc.

(R)-2-Amino-4-cyclohexylbutanoic acid

  • MDL number MFCD18252743
  • Cas Number : 728880-26-0
  • Molecular Weight : 185.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/728880-26-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/728880-26-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart