AMBH324A51F5 Ambeed, Inc.

4-Acetyl-L-phenylalanine Hydrochloride

  • MDL number MFCD12911919
  • Cas Number : 20299-31-4
  • Molecular Weight : 243.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/20299-31-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20299-31-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart