AMBH324A51F8 Ambeed, Inc.

((1S,3S,4S)-3-Bromo-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonic acid hydrate

  • MDL number MFCD00167452
  • Cas Number : 209736-59-4
  • Molecular Weight : 329.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/209736-59-4.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/209736-59-4.html
pictograms GHS05
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart