AMBH324A5203 Ambeed, Inc.

(R)-2-Amino-3-(7-methoxy-1H-indol-3-yl)propanoic acid

  • MDL number MFCD08437394
  • Cas Number : 808145-87-1
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/808145-87-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/808145-87-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart