AMBH324A5206 Ambeed, Inc.

trans-Benzyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD03427085
  • Cas Number : 95687-41-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/95687-41-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/95687-41-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart