AMBH324A520D Ambeed, Inc.

(S)-7-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD00144534
  • Cas Number : 128502-56-7
  • Molecular Weight : 193.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/128502-56-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/128502-56-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart