AMBH324A520E Ambeed, Inc.

N2-(1-Oxotetradecyl)-L-lysyl-L-alanyl-L-lysyl-L-alaninamide

  • MDL number N/A
  • Cas Number : 959610-24-3
  • Molecular Weight : 625.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/959610-24-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/959610-24-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart