AMBH324A5219 Ambeed, Inc.

Cabazitaxel

  • MDL number MFCD18827611
  • Cas Number : 183133-96-2
  • Molecular Weight : 835.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/183133-96-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/183133-96-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart