AMBH324A5223 Ambeed, Inc.

2-(Pyridin-4-yl)acetic acid hydrochloride

  • MDL number MFCD00012827
  • Cas Number : 6622-91-9
  • Molecular Weight : 173.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6622-91-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6622-91-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart