AMBH324A522E Ambeed, Inc.

(2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzyloxy)butanoic acid

  • MDL number MFCD00237033
  • Cas Number : 131545-63-6
  • Molecular Weight : 431.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131545-63-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/131545-63-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart