AMBH324A5244 Ambeed, Inc.

(1R,4R)-4-ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)

  • MDL number MFCD12911825
  • Cas Number : 139215-80-8
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/139215-80-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/139215-80-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart