AMBH324A5245 Ambeed, Inc.

(3R,4S)-tert-Butyl 3,4-dihydroxypyrrolidine-1-carboxylate

  • MDL number MFCD21919047
  • Cas Number : 186393-22-6
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/186393-22-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/186393-22-6.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart