AMBH324A524B Ambeed, Inc.

9-Amino minocycline hydrochloride

  • MDL number MFCD00919549
  • Cas Number : 149934-21-4
  • Molecular Weight : 508.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/149934-21-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/149934-21-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart