AMBH324A525C Ambeed, Inc.

(S)-1-tert-butyl 5-methyl 2-aminopentanedioate hydrochloride

  • MDL number MFCD02094380
  • Cas Number : 34582-33-7
  • Molecular Weight : 253.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/34582-33-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/34582-33-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart